Lab Pack Poly Drums, Eagle Mfg
Supplier: Eagle Manufacturing
1601 1600SL 1601MB 1601M 1601B 1601BLK 1601RED 1601MBLK 1601MBBR 1601BLKBG 1601MBBG 1601MBLKBG 1601M-BG2 1601MB-BG2 1601MBK-BG2 1601M-BRBG2 1601MB-BRBG2 1601MBK-BRBG2
56610-143EA 125.26 CAD
56610-143 76369-704 76369-706 56618-527 76502-650 76502-652 76502-654 76502-656 76502-658 76502-660 76502-662 76502-664 76502-666 76502-668 76502-670 76502-672 76502-674 76502-676
Lab Pack Poly Drums, Eagle Mfg
Barrels, Kegs, Drums
These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.
  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality -More Product Information