Dosing Spoons, Blue, Bürkle
Supplier: BURKLE INC
5378-4044 5378-4045 5378-4046 5378-4047 5378-4048 5378-4049 5378-4050 5378-4051 5378-4052 5378-4053 5378-4054 5378-4055 5378-4056 5378-4057 5378-4058 5378-4059 5378-4125
76501-486PK 60.66 CAD
76501-486 76501-488 76501-490 76501-492 76501-494 76501-496 76501-498 76501-500 76501-502 76501-504 76501-506 76501-508 76501-510 76501-512 76501-514 76501-516 76501-518
Dosing Spoons, Blue, Bürkle
Spoons
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly.
  • PS, blue
  • Optionally packaged individually and sterilised
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle. The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting intoMore Product Information