Evergreen Oro-Col™, Caplugs Evergreen
Supplier: Caplugs
240-3805-030
76499-448CT 469.97 CAD
76499-448
Evergreen Oro-Col™, Caplugs Evergreen
Funnels Safety Funnels
Oro-Col™ is an oral fluid collection system used primarily for drug abuse and hormone testing. Self-contained device for the direct collection and transport of oral fluid.
  • Simple in design is convenient to use
  • Consisting of a collection funnel and lid, calibrated vial and a screw cap
  • Collection funnel reduces the chance of contamination
  • All polypropylene construction