Press Lid Specimen Containers, Caplugs
Supplier: Caplugs
240-5701-06N 240-5702-06N 240-5703-06N 240-5704-06N 240-5705-06N 240-5706-06N
76499-424CT 191.54 CAD
76499-424 76499-426 76499-428 76499-430 76499-432 76499-434
Press Lid Specimen Containers, Caplugs
Sample Containers General Purpose Containers
Press lid specimen containers are designed for storing biological fluids and pathology specimens.
  • Press-close lids, and most other surface areas provide ample space for identification and other markings
  • All containers are precision molded from polyethylene which is highly resistant to most chemicals, including formaldehyde, most bases, and weak acids
  • Can withstand boiling and freezing
These multipurpose specimen containers are also useful for almost any miscellaneous storage needs, such as at laboratory or nursing stations.
Order Now
SPECIFICATIONS
Environmentally Preferable