PROPOR SG Filter Cartridges, Parker Bioscience
Supplier: PARKER HANNIFIN TE
ZCSGK-010C-PS ZCSGK-020C-PS ZCSGK-045C-PS ZCSG1-010C-PS ZCSG1-020C-PS ZCSG1-045C-PS ZCSG2-010C-PS ZCSG2-020C-PS ZCSG2-045C-PS ZCSG3-010C-PS ZCSG3-020C-PS ZCSG3-045C-PS ZCSGK-010C-PE ZCSGK-020C-PE ZCSGK-045C-PE ZCSG1-010C-PE ZCSG1-020C-PE ZCSG1-045C-PE ZCSG2-010C-PE ZCSG2-020C-PE ZCSG2-045C-PE ZCSG3-010C-PE ZCSG3-020C-PE ZCSG3-045C-PE ZCSGK-010D-PS ZCSGK-020D-PS ZCSGK-045D-PS ZCSG1-010D-PS ZCSG1-020D-PS ZCSG1-045D-PS ZCSG2-010D-PS ZCSG2-020D-PS ZCSG2-045D-PS ZCSG3-010D-PS ZCSG3-020D-PS ZCSG3-045D-PS ZCSGK-010D-PE ZCSGK-020D-PE ZCSGK-045D-PE ZCSG1-010D-PE ZCSG1-020D-PE ZCSG1-045D-PE ZCSG2-010D-PE ZCSG2-020D-PE ZCSG2-045D-PE ZCSG3-010D-PE ZCSG3-020D-PE ZCSG3-045D-PE ZCSGK-010E-PS ZCSGK-020E-PS ZCSGK-045E-PS ZCSG1-010E-PS ZCSG1-020E-PS ZCSG1-045E-PS ZCSG2-010E-PS ZCSG2-020E-PS ZCSG2-045E-PS ZCSG3-010E-PS ZCSG3-020E-PS ZCSG3-045E-PS ZCSGK-010E-PE ZCSGK-020E-PE ZCSGK-045E-PE ZCSG1-010E-PE ZCSG1-020E-PE ZCSG1-045E-PE ZCSG2-010E-PE ZCSG2-020E-PE ZCSG2-045E-PE ZCSG3-010E-PE ZCSG3-020E-PE ZCSG3-045E-PE
76497-508EA 954.34 CAD
76497-508 76497-510 76497-512 76497-514 76497-516 76497-518 76497-520 76497-522 76497-524 76497-526 76497-528 76497-530 76497-532 76497-534 76497-536 76497-538 76497-540 76497-542 76497-544 76497-546 76497-548 76497-550 76497-552 76497-554 76497-556 76497-558 76497-560 76497-562 76497-564 76497-566 76497-568 76497-570 76497-572 76497-574 76497-576 76497-578 76497-580 76497-582 76497-584 76497-586 76497-588 76497-590 76497-592 76497-594 76497-596 76497-598 76497-600 76497-602 76497-604 76497-606 76497-608 76497-610 76497-612 76497-614 76497-616 76497-618 76497-620 76497-622 76497-624 76497-626 76497-628 76497-630 76497-632 76497-634 76497-636 76497-638 76497-640 76497-642 76497-644 76497-646 76497-648 76497-650
PROPOR SG Filter Cartridges, Parker Bioscience
Filters Filter Cartridges
PROPOR SG sterilizing grade filters feature a microbially retentive polyethersulfone membrane for fast, reliable and cost-effective sterile filtration of pharmaceutical fluids.
  • Up to 3.5 times higher flow rates than competitive sterilizing grade filters
  • Fully validated and integrity testable membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use capsule format available with Pre-sterile version by Gamma-irradiation
The asymmetric pore structure and high voids volume of the PROPOR SG membrane allow high throughputs andMore Product Information