PROPOR HC Filter Cartridges, Parker Bioscience
Supplier: PARKER HANNIFIN TE
ZCHCK-620C-PS ZCHC1-620C-PS ZCHC2-620C-PS ZCHC3-620C-PS ZCHCK-620C-PE ZCHC1-620C-PE ZCHC2-620C-PE ZCHC3-620C-PE ZCHCK-620D-PS ZCHC1-620D-PS ZCHC2-620D-PS ZCHC3-620D-PS ZCHCK-620D-PE ZCHC1-620D-PE ZCHC2-620D-PE ZCHC3-620D-PE
76496-686EA 1071.15 CAD
76496-686 76496-688 76496-690 76496-692 76496-694 76496-696 76496-698 76496-700 76496-702 76496-704 76496-706 76496-708 76496-710 76496-712 76496-714 76496-716 76496-718 76496-720 76496-722 76496-724 76496-726 76496-728 76496-730 76496-732
PROPOR HC Filter Cartridges, Parker Bioscience
Filters Filter Cartridges
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
  • Optimized membrane configuration allows up to 10X the throughput compared to single layer membrane products
  • Integral prefilter layer can condense filter trains for greater processing economy
  • Incorporates a fully validated and integrity testable 0.2-micron membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use Capsule format available with pre-sterile version by Gamma-irradiation