Test Tubes, Caplugs
Supplier: Caplugs
214-2001-010 214-2002-010 214-2003-010 214-2009-010 214-2012-010 214-2023-010 214-2030-010 214-2065-01N 214-2068-G50 214-2070-010 214-2079-G60 214-2088-G50 214-2097-010 214-2099-G50 214-2131-010 214-2309-030 214-2313-030 214-2345-03N 214-2355-030 214-2367-030 214-2367-B30 214-2367-R30 214-2367-Y30 214-2380-030
76493-638BG 93.01 CAD
76493-638 76493-640 76493-642 76493-644 76493-646 76493-648 76493-650 76493-652 76493-654 76493-656 76493-658 76493-660 76493-662 76493-664 76493-666 76493-668 76493-670 76493-672 76493-674 76493-676 76493-678 76493-680 76493-682 76493-684
Test Tubes, Caplugs
Tubes Test Tubes
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
  • Provided with or without colors
  • Tubes are bulk packed, non-sterile
  • Polystyrene tubes withstand up to 8200×g
  • Polypropylene tubes withstand up to 10000×g
  • 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organicMore Product Information