VWR® Frio Temp Precision Thermometers
Supplier: VWR International
76491-672
76491-672CS 570.03 CAD
76491-672
VWR® Frio Temp Precision Thermometers
Thermometers Analog Thermometers
These glass thermometers can be used to take readings in freezers, refrigerators, incubators, ovens and blood banks.
  • Enviro safe non toxic and biodegradable liquid or red spirit liquid
  • Accurate temperature reading without sample contamination
  • Individual serial number
  • Wide choice
Accurate temperature reading is achieved by using the filled bottle (included), which mimics the sample. Insert the thermometer into the bottle and place near the samples for best results. PFA coating safely contains glass and liquid in case of breakage.
Certifications: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health