Borosil® Reusable Class A Serological Pipettes (Pipettes), Foxx Life Sciences®
Supplier: Foxx Life Sciences
7081P01A 7081P01D 7081P06A 7081P06D 7081P09A 7081P09D 7081P11A 7081P11D 7081P22A 7081P22D 7081P55A 7081P55D
76491-388CS 172.66 CAD
76491-388 76491-390 76491-392 76491-394 76491-396 76491-398 76491-400 76491-402 76491-404 76491-406 76491-408 76491-410
Borosil® Reusable Class A Serological Pipettes (Pipettes), Foxx Life Sciences®
Pipettes Serological Pipettes
Borosil® brand Pipettes are made of specially-selected, heavy wall, machine-drawn, borosilicate accurate, bore glass tubing. They have precision formed jets, and bevel ground ends that resist chipping.
  • Graduated to tip
  • Two top rings indicate to deliver (TD)
  • Permanent amber stain markings
The graduations are made in durable white enamel or amber stain that is resistant to laboratory reagents and repeat washing. All Pipettes are individually measured and then marked. The uniformity of bore ensures accuracy between any two readings and they areMore Product Information