Borosil® Volumetric Flasks with Interchangeable Solid Glass/Plastic Stopper, Foxx Life Sciences®
Supplier: Foxx Life Sciences
5645005A 5645005D 5645006A 5645006D 5645008A 5645008D 5645009A 5645009D 5645012A 5645012D 5645016A 5645016D 5645020A 5645020D 5645021A 5645021D 5645024A 5645024D 5645029A 5645029D 5645030A 5645030D 5655005A 5655005D 5655006A 5655006D 5655008A 5655008D 5655009A 5655009D 5655012A 5655012D 5655016A 5655016D 5655020A 5655020D 5655021A 5655021D 5655024A 5655024D 5655029A 5655029D 5655030A 5655030D
76491-140CS 161.72 CAD
76491-140 76491-142 76491-144 76491-146 76491-148 76491-150 76491-152 76491-154 76491-156 76491-158 76491-160 76491-162 76491-164 76491-166 76491-168 76491-170 76491-172 76491-174 76491-176 76491-178 76491-180 76491-182 76491-224 76491-226 76491-228 76491-230 76491-232 76491-234 76491-236 76491-238 76491-240 76491-242 76491-244 76491-246 76491-248 76491-250 76491-252 76491-254 76491-256 76491-258 76491-260 76491-262 76491-264 76491-266
Borosil® Volumetric Flasks with Interchangeable Solid Glass/Plastic Stopper, Foxx Life Sciences®
Flasks Volumetric Flasks
These Borosil® volumetric flasks with interchangeable PP stoppers are manufactured from 3.3 Borosilicate I.D. control tubing.
  • Type I Class A
  • Permanent amber markings
  • Calibrated TC (to Contain)
Each flask is individually calibrated with an automatic, computer controlled, calibration machine, providing a more accurate volume. Borosil’s volumetric flasks are designed to give the best combination of thermal shock resistance, physical strength and resistance to chemical attack. All volumetric flasks have permanent amber printing.
*Certified andMore Product Information