Borosil® Volumetric Pipette Bulbs Class A ASTM 969 USP Type I Individual Certification, Foxx Life Sciences®
Supplier: Foxx Life Sciences
7101001 7101002 7101003 7101004 7101005 7101007 7101008 7101009 7101010 7101011 7101012 7101016 7101105 7101205 7101705 7101706 7101707 7101708 7101709 7101711
76490-872CS 271.14 CAD
76490-872 76490-874 76490-876 76490-878 76490-880 76490-882 76490-884 76490-886 76490-888 76490-890 76490-892 76490-894 76490-896 76490-898 76490-900 76490-902 76490-904 76490-906 76490-908 76490-910 76490-912
Borosil® Volumetric Pipette Bulbs Class A ASTM 969 USP Type I Individual Certification, Foxx Life Sciences®
Pipettes Graduated Pipettes
Borosil Pipettes are made of heavy wall machine drawn 3.3 Borosilicate, bore glass tubing.
  • Calibrated to deliver
  • Each pipete coded
  • Round
The graduation marks are made of durable white enamel or amer stain resistant to laboratory reagents. All pipettes are individually measured and then marked. The uniformity of the bore ensures accuracy between any two readings.
All Borosil pipettes are color-coded at the top in accordance with ISO Standards to facilitate easy identification.
Certifications: Compliance with ASTM E 969-02.
Delivery information: Calibration certificate included.
Order Now
SPECIFICATIONS
Environmentally Preferable