Evergreen Serum Sample Cups, Caplugs
Supplier: Caplugs
127-0017-010 127-0017-020 127-0017-R10 127-0018-010 127-0018-020 127-0018-R10 127-0066-010 127-0066-R10 127-0080-010 127-0090-010 127-0093-010 127-0093-R10 127-1212-010 127-1225-010 127-1238-010 127-1240-010 127-1349-030
76490-446BG 151.38 CAD
76490-446 76490-448 76490-450 76490-452 76490-454 76490-456 76490-458 76490-460 76490-462 76490-464 76490-466 76490-468 76490-470 76490-472 76490-474 76490-476 76490-478 76490-480 76490-482 76490-484 76490-486
Evergreen Serum Sample Cups, Caplugs
Sample Containers Sample Cups for Analyzers
Serum sample cups are compatible with automated chemistry analyzers. Different serum cup materials accommodate to different chemicals.
  • Bulk with/without caps
  • Cups and caps come in a variety of colors
  • Polystyrene, polyethylene and polyproplene
Colors allow match to color-coding standards, controls, STATS and patient samples piggyback cups. Fit most high-volume analyzers. Polystyrene is transparent and polypropylene is unbreakable.
Order Now
SPECIFICATIONS
Environmentally Preferable