WB9000 Series Metal Wrist Band and Coil Cord Set, Transforming Technologies
Supplier: Transforming Technologies
WB0090A WB0090L WB0090M WB0090S WB9037A WB9037L WB9037M WB9037S WB9043A WB9043L WB9043M WB9043S
76490-236EA 29.37 CAD
76490-236 76490-238 76490-240 76490-242 76490-246 76490-248 76490-250 76490-252 76490-254 76490-256 76490-258 76490-260
WB9000 Series Metal Wrist Band and Coil Cord Set, Transforming Technologies
Grounders Wrist Grounders
High quality metal wrist strap set with an insulating outer surface and stainless steel inner surface.
  • Durable metal expansion wrist band
  • Available with a 6 or 12' coil cord
  • Multiple sizes are available, including an adjustable design
The WB9000 Series Metal Wrist Band and Coil Cord set is a premium band set featuring a high-quality metal wrist strap set with an insulating outer surface and stainless steel inner surface. The band is standard with 4 mm, machined male snap and provides a 360 degree conductive contact around the wrist, with maximum skinMore Product Information