ESD Rubber Table Mats, Textured, Transforming Technologies
Supplier: Transforming Technologies
MTT2436B MTT2436GN MTT2436GY MTT2436RB MTT2448B MTT2448GN MTT2448GY MTT2448RB MTT2460B MTT2460GN MTT2460GY MTT2460RB MTT2472B MTT2472GN MTT2472GY MTT2472RB MTT3036B MTT3036GN MTT3036GY MTT3036RB MTT3048B MTT3048GN MTT3048GY MTT3048RB MTT3060B MTT3060GN MTT3060GY MTT3060RB MTT3072B MTT3072GN MTT3072GY MTT3072RB MTT3648B MTT3648GN MTT3648GY MTT3648RB MTT3660B MTT3660GN MTT3660GY MTT3660RB MTT3672B MTT3672GN MTT3672GY MTT3672RB
76490-070EA 160.16 CAD
76490-070 76490-072 76490-074 76490-076 76490-078 76490-080 76490-082 76490-084 76490-086 76490-088 76490-090 76490-092 76490-094 76490-096 76490-098 76490-100 76490-102 76490-104 76490-106 76490-108 76490-110 76490-112 76490-114 76490-116 76490-118 76490-120 76490-122 76490-124 76490-126 76490-128 76490-130 76490-132 76490-134 76490-136 76490-138 76490-140 76490-142 76490-144 76490-146 76490-148 76490-150 76490-152 76490-154 76490-156
ESD Rubber Table Mats, Textured, Transforming Technologies
Surface Protectors Bench Protectors
The MT4000-Texture series ESD-safe 2-layer rubber matting is made of 100% co-extruded rubber.
  • Textured surface
  • Dissipative top layer, conductive bottom layer
  • 0.080" thick
  • Colors: Gray, green, blue, royal blue
The durable two layer mat is constructed with a textured dissipative top layer and a conductive bottom layer and offers excellent resistance to hot solders, soldering irons and most solvents. The MT-Texture series are available in 40' rolls in 24, 30, 36 and 48" widths. Pre cut mat sizes are available with grounding hardware and common pointMore Product Information