Static Field Meter, Transforming Technologies
Supplier: Transforming Technologies
EFM250
76489-642EA 2073.75 CAD
76489-642
Static Field Meter, Transforming Technologies
Electrical Meters Voltmeters
Perform static surveys and evaluate charge accumulation in electronic manufacturing and industrial operations such as converting, printing, packaging, and plastics processing.
  • LED guide: pulsing-beam rangefinder to consistently obtain correct readings
  • HOLD button captures transient readings
  • Measuring range: + 0 ~ 20 kV
The Static Field Meter is used to perform static surveys and evaluate static charge accumulation. The compact design and four button operation make it easy to operate. It can measure static charges up to 20 kV. Simply hold theMore Product Information