SP Bel-Art H-B Durac® Thermocouple Thermometer Probes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60901-1200
76486-054EA 207.26 CAD
76486-054 76486-056
SP Bel-Art H-B Durac® Thermocouple Thermometer Probes, Bel-Art Products, a part of SP
Temperature Probes
For use with H-B Durac® Calibrated Thermocouple Thermometers or others that accept K or J type probes.
  • For temperature measurement of liquids, semi-solids, or air/gasses
  • Measure wide temperature ranges
  • 15.24 cm (6") stainless steel probe
  • Model B60901-1300 perfect for use in dewars