VWR® Syringe Filters
Supplier: VWR International
76478-980 76478-982 76478-984 76478-986 76478-988 76478-990 76478-992 76478-994 76478-996 76478-998 76479-000 76479-002 76479-004 76479-006 76479-008 76479-010 76479-012 76479-014 76479-016 76479-018 76479-020 76479-022 76479-024 76479-026 76479-028 76479-030 76479-032 76479-034 76479-036 76479-038 76479-040 76479-042 76479-044 76479-046 76479-048 76479-050 76479-052 76479-054 76479-056 76479-058 76479-060
76478-980CS 4870.89 CAD
76478-980 76478-982 76478-984 76478-986 76478-988 76478-990 76478-992 76478-994 76478-996 76478-998 76479-000 76479-002 76479-004 76479-006 76479-008 76479-010 76479-012 76479-014 76479-016 76479-018 76479-020 76479-022 76479-024 76479-026 76479-028 76479-030 76479-032 76479-034 76479-036 76479-038 76479-040 76479-042 76479-044 76479-046 76479-048 76479-050 76479-052 76479-054 76479-056 76479-058 76479-060
VWR® Syringe Filters
Filters Syringe Filters
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are Ideal for sterile filtration
  • Nonsterile syringe filters are excellent for HPLC applications
  • Biosafe according to Class VI plastics tests
  • Available with Nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass fiber)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemicalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Autoclavable Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes Yes