VWR® Syringe Filters
Supplier: VWR International
76478-980 76478-982 76478-984 76478-986 76478-988 76478-990 76478-992 76478-994 76478-996 76478-998 76479-000 76479-002 76479-004 76479-006 76479-008 76479-010 76479-012 76479-014 76479-016 76479-018 76479-020 76479-022 76479-024 76479-026 76479-028 76479-030 76479-032 76479-034 76479-036 76479-038 76479-040 76479-042 76479-044 76479-046 76479-048 76479-050 76479-052 76479-054 76479-056 76479-058 76479-060
76478-980CS 4870.89 CAD
76478-980 76478-982 76478-984 76478-986 76478-988 76478-990 76478-992 76478-994 76478-996 76478-998 76479-000 76479-002 76479-004 76479-006 76479-008 76479-010 76479-012 76479-014 76479-016 76479-018 76479-020 76479-022 76479-024 76479-026 76479-028 76479-030 76479-032 76479-034 76479-036 76479-038 76479-040 76479-042 76479-044 76479-046 76479-048 76479-050 76479-052 76479-054 76479-056 76479-058 76479-060
VWR® Syringe Filters
Filters Syringe Filters
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
  • Sterile blister packed filters are Ideal for sterile filtration
  • Nonsterile syringe filters are excellent for HPLC applications
  • Biosafe according to Class VI plastics tests
  • Available with Nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass fiber)
  • Available with hydrophilic or hydrophobic PTFE
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
Nylon and PTFE membranes are ideal for sample preparation and small volume chemicalMore Product Information