MAGIO™ MS and MX Immersion Thermostats, Julabo
Supplier: JULABO
9032201.22 9032201.33 9033201.33
76477-120EA 12900.18 CAD
76477-120 76477-122 76477-124
MAGIO™ MS and MX Immersion Thermostats, Julabo
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The MAGIO™ series of bridge mounted circulators operate with a working temperature range of −50 to +300 °C* and a heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ immersion thermostats can be adapted to different tanks and applications.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
More Product Information