Bemcot® M-3II Static Dissipative Cleanroom Wipes, Transforming Technologies

Supplier: Transforming Technologies
WPBEMM-3II
76474-834EA 798.44 CAD
76474-834
Bemcot® M-3II Static Dissipative Cleanroom Wipes, Transforming Technologies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Static dissipative cleanroom ESD wipes are suitable for nearly all electronic manufacturing and optical cleaning applications.

  • Nonwoven, soft, and low-linting cotton wipe with excellent antistatic and heat resistance properties
  • Proprietary 100% cellulose filaments absorb 13 times its weight in liquid
  • No binders; safe to use with different solutions
  • Apertured surface promotes particle removal
  • 100% knitted polyester
  • No carbon or metal fibers
  • Less than 100 V turbo-charge

Wipes are designed for quick and easy wiping in cleanrooms and other workingMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR