Non-Marking ESD Heel Grounder, Transforming Technologies
Supplier: Transforming Technologies
HG1360NM-2
76474-660EA 21.19 CAD
76474-660
Non-Marking ESD Heel Grounder, Transforming Technologies
Grounders Heel and Toe Grounders
Non-marking stretch Hook and Loop heel grounders help keep controlled environments clean, and personnel grounded.
  • Made with ESD gray rubber that will not leave marks on floors
  • Stretchable hook and loop closure straps ensure a solid, comfortable fit
  • Reversible cup design allows for longer life
  • Made with a hi-vis blue strap that can be easily seen
The device is constructed from a non-marking and highly dissipative three-layer rubber. The reversible 1.5" wide rubber cup secures at the back and underneath the heel. The 7.5" length rubber provides enoughMore Product Information