PCRmax Alpha Cyclers, Cole Parmer
Supplier: Antyila Scientific
93945-04 93945-06 93945-08 93945-10 93945-12 93945-00 93945-02 93945-20 93945-22 93945-28
76474-120EA 43201.99 CAD
76474-120 76474-122 76474-124 76474-126 76474-128 76474-130 76474-132 76474-134 76474-136 76474-138
PCRmax Alpha Cyclers, Cole Parmer
Thermal Cyclers
Easily create, modify and access programs, saving time.
  • Flexible - your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
  • Auto Restart On Power Failure
  • Adjustable heated lid pressure
  • USB port output
These compact thermal cyclers deliver reproducible results every run - with little to no training needed. Features include a clear responsive touch screen, secure user-specificMore Product Information