Rapid Response™ Single Drug Test Strip (Urine), BTNX

Supplier: BTNX INC.
6MAM-1S5-50 BUP-1S5-100 COC-1S3-100 THC-1S13-100
76473-390KT 214.25 CAD
76473-390 76473-396 76473-398 76473-510
Rapid Response™ Single Drug Test Strip (Urine), BTNX
Drug Tests
The Rapid Response® Single Drug Test Strip Test is a rapid, qualitative test for the detection of a selected drug in human urine.

  • Sample: Urine
  • Format: Strip
  • Time to result: 5 minutes
  • Storage Condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Lateral flow immunoassay

Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line).

Single drug test strips can include BUP (10), COC (300), THC (50), and 6MAM (10).

Delivery information: Includes test strips, product insert.

Caution: This testMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR