Purifier® Axiom® Class II Type C1 Biosafety Cabinets, Labconco
Supplier: Labconco
304480001 304480011 304480021 304480031 304480041 304480051 304480071 304480101 304480111 304480121 304480131 304480141 304480151 304480171 304481001 304481011 304481021 304481031 304481041 304481051 304481071 304481101 304481111 304481121 304481131 304481141 304481151 304481171 304410001 304410011 304410021 304410031 304410041 304410051 304410071 304410101 304410111 304410121 304410131 304410141 304410151 304410171 304411001 304411011 304411021 304411031 304411041 304411051 304411071 304411101 304411111 304411121 304411131 304411141 304411151 304411171 304680001 304680011 304680021 304680031 304680041 304680051 304680071 304680101 304680111 304680121 304680131 304680141 304680151 304680171 304681001 304681011 304681021 304681031 304681041 304681051 304681071 304681101 304681111 304681121 304681131 304681141 304681151 304681171 304610001 304610011 304610021 304610031 304610041 304610051 304610071 304610101 304610111 304610121 304610131 304610141 304610151 304610171 304611001 304611011 304611021 304611031 304611041 304611051 304611071 304611101 304611111 304611121 304611131 304611141 304611151 304611171
76473-158EA 30833.7 CAD
76473-158 76473-160 76473-162 76473-164 76473-166 76473-168 76473-170 76473-172 76473-174 76473-176 76473-178 76473-180 76473-182 76473-184 76473-186 76473-188 76473-190 76473-192 76473-194 76473-196 76473-198 76473-200 76473-202 76473-204 76473-206 76473-208 76473-210 76473-212 76473-214 76473-216 76473-218 76473-220 76473-222 76473-224 76473-226 76473-228 76473-230 76473-232 76473-234 76473-236 76473-238 76473-240 76473-242 76473-244 76473-246 76473-248 76473-250 76473-252 76473-254 76473-256 76473-258 76473-260 76473-262 76473-264 76473-266 76473-268 76473-270 76473-272 76473-274 76473-276 76473-278 76473-280 76473-282 76473-284 76473-286 76473-288 76473-290 76473-292 76473-294 76473-296 76473-298 76473-300 76473-302 76473-304 76473-306 76473-308 76473-310 76473-312 76473-314 76473-316 76473-318 76473-320 76473-322 76473-324 76473-326 76473-328 76473-330 76473-332 76473-334 76473-336 76473-338 76473-340 76473-342 76473-344 76473-346 76473-348 76473-350 76473-352 76473-354 76473-356 76473-358 76473-360 76473-362 76473-364 76473-366 76473-368 76473-370 76473-372 76473-374 76473-376 76473-378 76473-380
Purifier® Axiom® Class II Type C1 Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Flexible BSCs can be operated in Type A or B mode and changed as application needs change.
  • NSF Listed
  • Only Type C1 on the market
  • Maximum safety and flexibility
The Axiom Class II Type C1 maximizes safety and flexibility as the first biosafety cabinet that can be used in a recirculating Type A mode for standard microbiological work or can be connected to an exhaust system to function in Type B mode for handling hazardous chemicals. The unique design includes the Chem-Zone™, providing chemical safety with a large, clearly defined area within theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable