Bottle Brush, Micronova
Supplier: Micronova
BRWH-01 BRWH-01B
89108-046CS 899.04 CAD
89108-046 76472-546
Bottle Brush, Micronova
Brushes
Multipurpose brushes with rounded polyester bristles for versatile cleaning of bottles and various equipment in labs, cleanrooms and production facilities. Handles are polypropylene. Brushes are BPA free.
  • Multipurpose, color-coded plastic bottle brushes
  • Durable polyester bristles
  • Stiff bristles for aggressive cleaning of bottles and equipment
  • Soft bristles for delicate cleaning and smaller openings
  • 12 or 16-inch easy grip, plastic handles
  • Made from FDA approved, BPA free base resins
  • Available in multiple colors
The Micronova M-Zone™Bottle BrushesMore Product Information