Bedside Carts, TrippNT
Supplier: TrippNT
53401 53406 53407 53549
76471-892EA 1541.03 CAD
76471-892 76471-894 76471-896 76471-898 76471-900 76471-902 76471-904 76471-906 76471-908 76471-910 76471-912 76471-914 76471-916
Bedside Carts, TrippNT
Trolleys
TrippNT's Bedside Cart offers total locking mobile storage.
  • 2 locking casters, 2 swivel casters, all locking drawer and bulk storage area
  • Built-in recessed tray in the top of the cart
  • Corrosion-resistant, non-metal HDPE construction
  • Designed to secures personal effects during transit and stationary use
  • 100% total replacement lifetime guarantee/made in USA
TrippNT's Bedside Cart is used to protect and store patient’s belongings. The top of the cart is inset to create a tray to secures items such as: phones, drinks, and other possessions that need to beMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable