VWR® ETO Sterilized Alliance Sleeves, Made in USA
Supplier: VWR International
76470-704
76470-704CS 2352.4 CAD
76470-704
VWR® ETO Sterilized Alliance Sleeves, Made in USA
Disposable Garments
VWR® Made in USA Sterile Sleeves are an ideal option for single-use needs. Made with the specially formulated Alliance breathable microporous fabric.
  • Individually bagged
  • ETO sterilized
  • Made with Alliance breathable microporous fabric
  • Made in USA
A key part of the VWR® Made in USA PPE line, these sterile cleanroom sleeves are 21" in length to provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outerMore Product Information