SP Bel-Art® Sterileware® Upright Handle Dippers or Ladles, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36775-0050 F36775-1050
76469-928PK 349.76 CAD
76469-928 76469-930
SP Bel-Art® Sterileware® Upright Handle Dippers or Ladles, Bel-Art Products, a part of SP
Samplers Sample Dippers
Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams.
  • Upright handle allows for sampling from above container
  • Curved handle end provides secure withdrawal of sample
  • Fits into 2 in. openings such as a 45-gallon drum bunghole
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • White high density polyethylene (HDPE), or blue polypropylene (PP) ladles heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped forMore Product Information