PCR Tube and Cap Strips, Globe Scientific
Supplier: Globe Scientific
PCR-02DC PCR-02FC PCR-02S PCR-DL-01F PCR-DL-01FW PCR-DL-02F PCR-QS-02D PCR-QS-02F PCR-STR-01F PCR-STR-01FW PCR-STR-02D PCR-STR-02F
76468-966BX 55.66 CAD
76468-966 76468-968 76468-970 76468-972 76468-974 76468-976 76468-978 76468-980 76468-982 76468-984 76468-986 76468-988
PCR Tube and Cap Strips, Globe Scientific
Tubes PCR Tubes
The Diamond® PCR Strip tubes and caps offering includes DiamondLink™ strips which feature extremely strong links between wells for ease of handling and QuickSnap™ strips that easily snap apart for customization into smaller strips or use in multiple analyses.
  • Ultra-thin wall design (only 0.2 mm thick) for optimal temperature transfer
  • Certified free of detectable DNase, RNase, Human DNA, PCR inhibitors and tested Pyrogen-free
  • DiamondLink™ strips include 45° nested attached individual caps and strong links between tubes for easy handling
  • QuickSnap™ also
More Product Information