Hughes Gravity Fed Eyewash Insulated and Heated Jackets, Justrite®
Supplier: Justrite
10GFEW-BLKT 10GFEW-BLKT-HT 16GFEW-BLKT 16GFEW-BLKT-HT
76462-632EA 955.88 CAD
76462-632 76462-634 76462-636 76462-638
Hughes Gravity Fed Eyewash Insulated and Heated Jackets, Justrite®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Insulated and heated jackets can be quickly installed on the OptiWash™, using hook and loop fasteners and drawstrings.
  • Jackets offer both frost and heat protection of the OptiWash™
  • Jackets are easily installed on the OptiWash™, using hook and loop fasteners and drawstrings
  • Eyewash easily activated by pulling orange activation arm down to the open position
  • Heated jackets include 10' power cord with UL listed GFCI plug
  • 120 V power supply required to run heated jacket (60 W power rating); heated jackets only
  • UL approved thermostat to control temperature, and
More Product Information