Brady® Thermal Transfer 1 Core R6000 Halogen-Free Series Printer Ribbon
Supplier: Brady Worldwide
R6602 R6007
76460-978RL 387.36 CAD
76460-978 76460-980
Brady® Thermal Transfer 1 Core R6000 Halogen-Free Series Printer Ribbon
Printer Ribbons
This ink-side-out ribbon features an open 1" core for compatibility with a wide variety of thermal transfer printers.
  • Halogen-free
  • Heat-resistant
  • Scratch-resistant
  • Solvent-resistant
One-color ribbon for printing a single color onto a label. R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material. Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 Permanent polyester from Brady®. Combine Brady labelMore Product Information