LABOPORT™ N820G and N840G Vacuum Diaphragm Pumps, KNF
Supplier: KNF Neuberger Inc.
317121/313453 317123/315995
76460-434EA 5027.01 CAD
76460-434 76460-436
LABOPORT™ N820G and N840G Vacuum Diaphragm Pumps, KNF
Pumps Vacuum Diaphragm Pumps
Ideal for a range of lab applications such as rotary evaporation, degassing and vacuum ovens.
  • TFM™ PTFE head, FFPM valves and PTFE-coated diaphragm are ideal for aggressive/corrosive gases and vapors
  • 100% oil-free operation for pure transferring, evacuation and compression of gases
  • Integrated gas ballast valve, and rotational speed control tailors the pump to specific needs
  • 3 color status display indicates in-operation/stand-by/error
  • Environmentally friendly
The pump's small footprint saves valued bench space and its integrated speed control enablesMore Product Information