SP Wilmad-LabGlass Rotors and Caps for Bruker® MAS Probes, SP Industries
Supplier: SP Industries
JK-601-4181 JK-602-4181 JK-603-4181 JK-604-4181 WP-501-3180 WP-501-3180-SET1 WP-501-4180 WP-501-4180-SET-1 WP-501-4180-SET-2 WP-501-4180-SET-5 WP-501-4181 WP-501-7180 WP-501-7180-SET-2 WP-601-4181 WP-602-3182 WP-603-3181
76460-034EA 224.29 CAD
76460-034 76460-036 76460-038 76460-040 76460-042 76460-044 76460-046 76460-048 76460-050 76460-282 76460-284 76460-286 76460-288 76460-290 76460-292 76460-294 76460-296 76460-298 76460-300 76460-302 76460-304 76460-306 76460-308 76460-310 76460-312 76460-314 76460-316 76460-318 76460-320 76460-322
SP Wilmad-LabGlass Rotors and Caps for Bruker® MAS Probes, SP Industries
Centrifuges Rotors
MAS-NMR rotor bodies are manufactured from the highest quality Zirconia, Kel-F, Torlon® and Vespel® providing the ultimate solution for analysis of solid samples.
  • MAS rotors and caps are 100% compatible with most solid state NMR spectrometers
  • Thoroughly inspected before and after the precision machining process to ensure there are no material irregularities
  • Spin testing is performed to the highest specified spinning speed, assuring performance without overspinning the rotor
  • Spinning speeds of up to 12 kHz for 7 mm O.D. rotors
  • Some caps are fitted with O-
More Product Information