Avantor® ACE® UltraCore Biphenyl, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
CORE-35D-521 CORE-35D-530 CORE-35D-546 CORE-35D-1021 CORE-35D-1030 CORE-35D-1046 CORE-35D-1521 CORE-35D-1530 CORE-35D-1546 CORE-35D-21GD CORE-35D-30GD CORE-35D-46GD
76458-016EA 1411.52 CAD
76458-016 76458-018 76458-020 76458-022 76458-024 76458-026 76458-028 76458-030 76458-032 76458-034 76458-036 76458-038
Avantor® ACE® UltraCore Biphenyl, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore Biphenyl are high performing solid-core (core-shell) particle columns, designed for a range of chromatographic applications and is especially well suited for retention of aromatic compounds that elute early on alkyl phases.
  • High purity, base deactivated silica with excellent reproducibility
  • Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
  • Alternative selectivity to alkyl phases and traditional phenyl phases
These stainless steel columns are available in a variety of dimensions and are highlyMore Product Information