Avantor® ACE® C18 UltraCore BIO, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
BIO-250-521G BIO-250-0521 BIO-250-1021 BIO-250-1521 BIO-250-530G BIO-250-0530 BIO-250-1030 BIO-250-1530 BIO-250-546G BIO-250-0546 BIO-250-1046 BIO-250-1546 BIO-350-521G BIO-350-1021 BIO-350-1521 BIO-350-530G BIO-350-0530 BIO-350-1030 BIO-350-1530 BIO-350-546G BIO-350-0546 BIO-350-1046 BIO-350-1546
76457-846EA 1367.38 CAD
76457-846 76457-852 76457-858 76457-864 76457-870 76457-876 76457-882 76457-888 76457-894 76457-900 76457-906 76457-912 76457-918 76457-924 76457-930 76457-936 76457-942 76457-948 76457-954 76457-960 76457-966 76457-972 76457-978 76457-984
Avantor® ACE® C18 UltraCore BIO, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore BIO C18 are high performing solid-core (core-shell) particle columns designed for a wide range of biomolecule applications. With both 300 and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of more hydrophilic proteins over 5 kDa molecular weight.
  • Wide pore, solid-core combination for excellent peak shape and resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Good choice for fast analysis of polypeptides
These stainless steelMore Product Information