Avantor® ACE® C4 UltraCore BIO, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
BIO-251-521G BIO-251-0521 BIO-251-1021 BIO-251-1521 BIO-251-530G BIO-251-0530 BIO-251-1030 BIO-251-1530 BIO-251-546G BIO-251-0546 BIO-251-1046 BIO-251-1546 BIO-351-521G BIO-351-0521 BIO-351-1021 BIO-351-1521 BIO-351-530G BIO-351-0530 BIO-351-1030 BIO-351-1530 BIO-351-546G BIO-351-0546 BIO-351-1046 BIO-351-1546
76457-848EA 1367.38 CAD
76457-848 76457-854 76457-860 76457-866 76457-872 76457-878 76457-884 76457-890 76457-896 76457-902 76457-908 76457-914 76457-920 76457-926 76457-932 76457-938 76457-944 76457-950 76457-956 76457-962 76457-968 76457-974 76457-980 76457-986
Avantor® ACE® C4 UltraCore BIO, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® UltraCore BIO C4 are high performing solid-core (core-shell) particle columns, designed for a range of biomolecule applications. With both 300 and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of hydrophobic proteins over 5 kDa molecular weight.
  • Wide pore, solid-core combination for excellent peak shape and resolution of large molecules
  • High purity, base deactivated silica with excellent reproducibility
  • Butyl ligand well established in RP protein applications
These stainless steelMore Product Information