I2456 Cut Resistant Impact Gloves, Wells Lamont
Supplier: Wells Lamont
I2456S I2456M I2456L I2456XL I2456XXL
76451-966CS 2104.78 CAD
76451-966 76451-968 76451-970 76451-972 76451-974
I2456 Cut Resistant Impact Gloves, Wells Lamont
Gloves
Lightweight glove delivers great cut protection.
  • Hi-Vis
  • TPR impact protection
  • Reinforced thumb crotch
It provides protection for a wide range of applications with impact and cut hazards. The lightweight liner also provides great dexterity and comfort along with the padded palm for added protection. The TPR on the back-of-hand and the fingers is great for deflecting and absorbing impact as well. A sandy nitrile coating on the palm provides superior grip in oily applications.
Certifications: ANSI cut A5, abrasion level 5,More Product Information