GMP Covers, 1422A Tyvek®, Autoclavable, Cylindrical Shaped, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
COVTVK0716E COVTVK1048E COVTVK1416 COVTVK1416E COVTVK1460 COVTVK1460E COVTVK1613 COVTVK1613E COVTVK2009E COVTVK2048 COVTVK2048E COVTVK3012E COVTVK3260 COVTVK3260E COVTVK4410E COVTVK4836 COVTVK4836E COVTVK5060E COVTVK6036E COVTVK7248WS8T
76451-738CS 9098.43 CAD
76451-738 76451-740 76451-742 76451-744 76451-746 76451-748 76451-750 76451-752 76451-754 76451-756 76451-758 76451-760 76451-762 76451-764 76451-766 76451-768 76451-770 76451-772 76451-776 76451-778
GMP Covers, 1422A Tyvek®, Autoclavable, Cylindrical Shaped, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for steam sterilization; or use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Individually poly bagged and labeled
  • Steam Sterilization Indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
GMP Tyvek Covers provide a convenient, time-saving method of wrappingMore Product Information