Tyvek® 1073B Self-Sealing Autoclave Bags with Steam Indicator, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
COVMGTVK114SS COVMGTVK36SS COVMGTVK336SS COVMGTVK525SS COVMGTVK68SS COVMGTVK616SS COVMGTVK622SS COVMGTVK634SS COVMGTVK89SS COVMGTVK812SS COVMGTVK814SS COVMGTVK830SS COVMGTVK1012SS COVMGTVK1020SS COVMGTVK1025SS COVMGTVK1212SS COVMGTVK1216SS COVMGTVK1218SS COVMGTVK1224SS COVMGTVK1232SS COVMGTVK1234SS COVMGTVK1422SS COVMGTVK1428SS COVMGTVK1442SS COVMGTVK1518SS COVMGTVK1624SS COVMGTVK1636SS COVMGTVK1644SS COVMGTVK1822SS COVMGTVK1824SS COVMGTVK1826SS COVMGTVK1830SS COVMGTVK1840SS COVMGTVK1868SS COVMGTVK2020SS COVMGTVK2032SS COVMGTVK2330SS COVMGTVK2424SS COVMGTVK2444SS COVMGTVK2532SS COVMGTVK2537SS COVMGTVK2628SS COVMGTVK2630SS COVMGTVK3030SS COVMGTVK3022SS COVMGTVK3032SS COVMGTVK3234SS COVMGTVK3434SS COVMGTVK3440SS COVMGTVK3636SS COVMGTVK3642SS COVMGTVK3648SS COVMGTVK3660SS COVMGTVK3840SS COVMGTVK3848SS COVMGTVK4048SS COVMGTVK4246SS COVMGTVK4828SS COVMGTVK4848SS COVMGTVK4864SS
76450-632CS 993.79 CAD
76450-632 76450-634 76450-636 76450-638 76450-640 76450-642 76450-644 76450-646 76450-648 76450-650 76450-652 76450-654 76450-656 76450-658 76450-660 76450-662 76450-664 76450-666 76450-668 76450-670 76450-672 76450-674 76450-676 76450-678 76450-680 76450-682 76450-684 76450-686 76450-688 76450-690 76450-692 76450-694 76450-696 76450-698 76450-700 76450-702 76450-704 76450-706 76450-708 76450-710 76450-712 76450-714 76450-716 76450-718 76450-720 76450-722 76450-724 76450-726 76450-728 76450-730 76450-732 76450-734 76450-736 76450-738 76450-740 76450-742 76450-744 76450-746 76450-748 76450-750
Tyvek® 1073B Self-Sealing Autoclave Bags with Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-Sealing 1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges. No need to use or validate a heat sealing machine.
  • Manufactured and packaged in a controlled cleanroom environment
  • Convenient Self-sealing closure system
  • Steam sterilization indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant, hydrophobic, lint-free
  • 100% Medical Grade 1073B Tyvek® permits fast steam penetration
  • Excellent impact resistance, exceptional
More Product Information