Mini-Muff Closures with Tyvek® Drawstring Closure, DuPont™, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFT2A05D MFT2A07D MFT2A09D MFT2A12D MFT2A16D
76450-408CS 6517.5 CAD
76450-408 76450-410 76450-412 76450-414 76450-416
Mini-Muff Closures with Tyvek® Drawstring Closure, DuPont™, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closuresMore Product Information