BHD Autoclavable Muff Covers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFNWB04 MFNWB05 MFNWB06 MFNWB07 MFNWB08 MFNWB09 MFNWB12 MFNWB16 MFNWB18
76450-346CS 2346.3 CAD
76450-346 76450-348 76450-350 76450-352 76450-354 76450-356 76450-358 76450-360 76450-362
BHD Autoclavable Muff Covers, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. BHD Muffs provide a convenient means to cover flasks,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable