VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Supplier: VWR International
76449-278
76449-278CS 48.41 CAD
76449-278
VWR® Aluminum Sealing Strips for 8-Strip PCR Tubes
Seals Microplate Seals
Easier to apply and remove as compared to standard molded PCR strip caps.
  • Ideal for PCR
  • User-friendly alternative to strip-caps; no sore thumbs and fingers guaranteed
  • Seals a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Certified DNase- and RNase-free
  • 2.0 mil pierceable foil
  • Recommended temperature range from −20 to +120 °C
6 strips are precut on a continuous liner for ease of use.