Glassware washer accessories, FlaskScrubber® and SteamScrubber®
Supplier: Labconco
4573000 4573100 4573400 4573500 4573600 4573700 4573800 4573900 4574200 4574600 4575700 4574700 4574800 4574900 4575200 4575300 4575400 4575500 4574400 4574500 4587520 4595120 4679700 4653500 4653300 4678000 4643000 4669540 4679300 4679301 4668501 4668601 4668901 4668500 4668600 4668700 4668800 4668900 4669000 4575800 4678400 4678401 4596110 4596111 4681300 4681301
76449-150EA 361.82 CAD
76449-150 76449-152 76449-154 76449-156 76449-158 76449-160 76449-162 76449-164 76449-166 76449-168 76449-170 76449-172 76449-174 76449-176 76449-178 76449-180 76449-182 76449-184 76449-186 76449-188 76449-190 76449-192 76449-194 76449-196 76449-198 76449-200 76449-202 76449-204 76449-206 76449-208 76449-210 76449-212 76449-214 76449-216 76449-218 76449-220 76449-222 76449-224 76449-226 76449-228 76449-230 76449-232 76449-234 76449-236 76449-238 76449-240 76449-242
Glassware washer accessories, FlaskScrubber® and SteamScrubber®
Glassware Washers
There is a wide selection of accessories to customize your glassware washer to your specific needs and applications. These racks and accessories are for use with Labconco FlaskScrubber®, SteamScrubber®, FlaskScrubber® Vantage glassware washers. Racks and inserts are stainless steel unless otherwise stated.
  • Versatile inserts and specialized wash accessories
  • Additional, interchangeable racks for upper and lower positions
  • Optional accessories to make the most of your Labconco glassware washer including printers and automatic detergent dispensers
Order Now
SPECIFICATIONS
Environmentally Preferable