VWR® Polystyrene Assay Plates, 96- and 384-Well
Supplier: VWR International
76221-764 76446-954 76446-956 76446-958 76446-960 76446-962 76446-964 76446-970 76446-972 76446-974 76446-980 76446-982 76446-984 76446-986 76446-988 76446-990 76446-992 76446-994 76446-996 76446-998 76447-002 76447-004 76447-006 76447-008 76447-010 76447-012
76221-764CS 547.35 CAD
76221-764 76446-954 76446-956 76446-958 76446-960 76446-962 76446-964 76446-970 76446-972 76446-974 76446-980 76446-982 76446-984 76446-986 76446-988 76446-990 76446-992 76446-994 76446-996 76446-998 76447-002 76447-004 76447-006 76447-008 76447-010 76447-012
VWR® Polystyrene Assay Plates, 96- and 384-Well
Microplates HTS Microplates
Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
  • Flat bottom ideal for microscopic applications
  • V-bottom minimising residual liquid
  • U-bottom for optimising sample/cell collection
VWR® assay plates are available in clear, white or black polystyrene, with solid or clear bottom.
The bottom of the clear bottom plates is just 0.4 mm thick and extremely flat (±0.1 mm tolerance), ensuring high measurement precision and elimination of reading errors when performing cell-based assays usingMore Product Information