VWR® T Balances
Supplier: VWR International
76300-056 76300-070 76300-072 76300-074 76300-076 76300-078 76300-080 76300-082 76446-794 76446-796 76446-798 76446-800 76446-802 76446-804 76446-806 76446-808
76300-056EA 10700.19 CAD
76300-056 76300-070 76300-072 76300-074 76300-076 76300-078 76300-080 76300-082 76446-794 76446-796 76446-798 76446-800 76446-802 76446-804 76446-806 76446-808
VWR® T Balances
Balances and Scales
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
  • 2× RS232, USB (A and B), ethernet and Wi-Fi communication interfaces
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude ofMore Product Information