HALO® ES-CN, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
92122-104 92123-104 92124-104 95122-104 95123-104 95124-104 91225-404 91225-604 91225-704 91226-404 91226-604 91226-704 92120-404 92120-604 92120-704 92121-404 92121-604 92121-704 92121-904 92122-204 92122-304 92122-404 92122-504 92122-604 92122-704 92122-904 92123-204 92123-304 92123-404 92123-504 92123-604 92123-704 92123-904 92124-204 92124-304 92124-404 92124-504 92124-604 92124-704 92124-904 95120-404 95120-604 95120-704 95122-204 95122-304 95122-404 95122-504 95122-604 95122-704 95123-204 95123-304 95123-404 95123-504 95123-604 95123-704 95124-204 95124-304 95124-404 95124-504 95124-604 95124-704 95120-904 95122-904 95123-904 95124-904
76443-370PK 1124.96 CAD
76443-370 76443-374 76443-378 76443-540 76443-544 76443-548 76443-698 76443-702 76443-706 76443-710 76443-714 76443-718 76443-722 76443-726 76443-730 76443-734 76443-738 76443-742 76443-746 76443-750 76443-754 76443-758 76443-762 76443-766 76443-770 76443-774 76443-778 76443-782 76443-786 76443-790 76443-794 76443-798 76443-802 76443-806 76443-810 76443-814 76444-218 76444-222 76444-226 76444-230 76444-234 76444-238 76444-242 76444-246 76444-250 76444-254 76444-258 76444-262 76444-266 76444-270 76444-274 76444-278 76444-282 76444-286 76444-290 76444-294 76444-298 76444-302 76444-306 76444-310 76444-314 76444-318 76444-322 76444-326 76444-330 76444-334 76444-338 76445-288 76445-292 76445-296 76445-300 76445-306 76445-310 76445-314 76445-318 76445-322 76445-328 76445-332 76445-336 76445-340 76445-344 76445-348 76445-354 76445-358 76445-362 76445-366 76445-370 76445-374 76445-380 76445-384 76445-388 76445-392 76445-396 76445-400 76446-738 76446-744 76446-746 76446-748 76446-750 76446-752
HALO® ES-CN, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS ES-CN Peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast- and ultrahigh-resolution separations of peptides and polypeptides up to 20 kDa. HALO® peptide columns offer comparable peak capacity to sub-2 μm non-core columns at 40 to 50% back pressure (2.7 μm). HALO® 160 Å peptide columns are optimized for high performance in HPLC, UHPLC and LCMS peptide applications.
  • Ideal for tryptic digests, post-translational modifications (PTMs), polypeptides
  • HALO® Peptide ES-CN offers improved retention for
More Product Information