HALO® Diphenyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
93412-126 93414-126 93412-426 93412-626 93412-726 93414-426 93414-626 93414-726
76443-526PK 1124.96 CAD
76443-526 76443-536 76445-140 76445-150 76445-156 76445-202 76445-212 76445-218
HALO® Diphenyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS Diphenyl 400 Å protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high-resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 400 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
  • Ideal for large protein analysis such as mAbs and polypeptides
  • Well suited for high speed protein separations
  • Exceptional recovery
More Product Information