HALO® C18, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
91812-102 91813-102 92812-102 92813-102 92814-102 95812-102 95813-102 95814-102 91812-202 91812-302 91812-402 91812-502 91812-602 91812-702 91812-902 91813-202 91813-302 91813-402 91813-502 91813-602 91813-702 91813-902 92810-402 92810-602 92810-702 92811-402 92811-602 92811-702 92812-202 92812-302 92812-402 92812-502 92812-602 92812-702 92812-902 92813-202 92813-302 92813-402 92813-502 92813-602 92813-702 92813-902 92814-202 92814-302 92814-402 92814-502 92814-602 92814-702 92814-902 95810-402 95810-602 95810-702 95810-902 95812-202 95812-302 95812-402 95812-502 95812-602 95812-702 95812-902 95813-202 95813-302 95813-402 95813-502 95813-602 95813-702 95813-902 95814-202 95814-302 95814-402 95814-502 95814-602 95814-702 95814-902 98215-402 98215-602 98215-702 98216-402 98216-602 98216-702 9281X-402 9281X-602 9281X-702
76443-318PK 1124.96 CAD
76443-318 76443-338 76443-428 76443-460 76443-492 76443-552 76443-572 76443-592 76443-818 76443-838 76443-858 76443-878 76443-898 76443-918 76443-938 76443-958 76443-978 76443-998 76444-018 76444-038 76444-058 76444-078 76444-512 76444-528 76444-544 76444-560 76444-576 76444-596 76444-616 76444-636 76444-652 76444-668 76444-688 76444-708 76444-728 76444-748 76444-764 76444-780 76444-796 76444-816 76444-836 76444-856 76444-876 76444-892 76444-908 76444-924 76444-944 76444-964 76444-984 76445-004 76445-022 76445-042 76445-406 76445-426 76445-446 76445-466 76445-486 76445-506 76445-526 76445-546 76445-566 76445-586 76445-606 76445-626 76445-646 76445-666 76445-686 76445-706 76445-726 76445-746 76445-766 76445-786 76445-806 76445-826 76445-846 76445-866 76445-886 76445-906 76445-926 76445-946 76445-966 76445-986 76446-006 76446-026 76446-136 76446-156 76446-176 76446-196 76446-216 76446-236 76446-256 76446-276 76446-296 76446-316 76446-336 76446-356 76446-376 76446-396 76446-416 76446-436 76446-456 76446-476 76446-496 76446-516 76446-536 76446-556 76446-576 76446-596 76446-616 76446-636 76446-656 76446-676 76446-696 76446-716 76537-470 76537-472 76537-474
HALO® C18, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® C18 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å C18 columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Universal phase for acids, bases and neutral solutes with a wide range of hydrophobicities
  • Excellent stability at low to mid mobile phase pH
  • Widely published and most utilized stationary phase
  • Particle sizes: 2, 2.
More Product Information