HALO® HILIC, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
91812-101 91813-101 92812-101 92813-101 92814-101 95812-101 95813-101 95814-101 91812-201 91812-301 91812-401 91812-501 91812-601 91812-701 91812-901 91813-201 91813-301 91813-401 91813-501 91813-601 91813-701 91813-901 92810-401 92810-601 92810-701 92811-401 92811-601 92811-701 92812-201 92812-301 92812-401 92812-501 92812-601 92812-701 92812-901 92813-201 92813-301 92813-401 92813-501 92813-601 92813-701 92813-901 92814-201 92814-301 92814-401 92814-501 92814-601 92814-701 92814-901 95810-401 95810-601 95810-701 95810-901 95812-201 95812-301 95812-401 95812-501 95812-601 95812-701 95812-901 95813-201 95813-301 95813-401 95813-501 95813-601 95813-701 95813-901 95814-201 95814-301 95814-401 95814-501 95814-601 95814-701 95814-901 98215-401 98215-601 98215-701 98216-401 98216-601 98216-701
76443-316PK 1124.96 CAD
76443-316 76443-336 76443-426 76443-458 76443-490 76443-550 76443-570 76443-590 76443-816 76443-836 76443-856 76443-876 76443-896 76443-916 76443-936 76443-956 76443-976 76443-996 76444-016 76444-036 76444-056 76444-076 76444-510 76444-526 76444-542 76444-558 76444-574 76444-594 76444-614 76444-634 76444-650 76444-666 76444-686 76444-706 76444-726 76444-746 76444-762 76444-778 76444-794 76444-814 76444-834 76444-854 76444-874 76444-890 76444-906 76444-922 76444-942 76444-962 76444-982 76445-002 76445-020 76445-040 76445-404 76445-424 76445-444 76445-464 76445-484 76445-504 76445-524 76445-544 76445-564 76445-584 76445-604 76445-624 76445-644 76445-664 76445-684 76445-704 76445-724 76445-744 76445-764 76445-784 76445-804 76445-824 76445-844 76445-864 76445-884 76445-904 76445-924 76445-944 76445-964 76445-984 76446-004 76446-024 76446-134 76446-154 76446-174 76446-194 76446-214 76446-234 76446-254 76446-274 76446-294 76446-314 76446-334 76446-354 76446-374 76446-394 76446-414 76446-434 76446-454 76446-474 76446-494 76446-514 76446-534 76446-554 76446-574 76446-594 76446-614 76446-634 76446-654 76446-674 76446-694 76446-714
HALO® HILIC, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for polar analytes
  • Alternate mode to reversed-phase modes
  • Can be used in HILIC and normal-phase modes
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')