HALO® AQ-C18, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
91812-122 91813-122 92812-122 92813-122 92814-122 95812-122 95813-122 95814-122 91812-222 91812-322 91812-422 91812-522 91812-622 91812-722 91812-922 91813-222 91813-322 91813-422 91813-522 91813-622 91813-722 91813-922 92810-422 92810-622 92810-722 92811-422 92811-622 92811-722 92812-222 92812-322 92812-422 92812-522 92812-622 92812-722 92812-922 92813-222 92813-322 92813-422 92813-522 92813-622 92813-722 92813-922 92814-222 92814-322 92814-422 92814-522 92814-622 92814-722 92814-922 95810-422 95810-622 95810-722 95810-922 95812-222 95812-322 95812-422 95812-522 95812-622 95812-722 95812-922 95813-222 95813-322 95813-422 95813-522 95813-622 95813-722 95813-922 95814-222 95814-322 95814-422 95814-522 95814-622 95814-722 95814-922 98215-422 98215-622 98215-722 98216-422 98216-622 98216-722
76443-334PK 1124.96 CAD
76443-334 76443-354 76443-444 76443-476 76443-508 76443-568 76443-588 76443-608 76443-834 76443-854 76443-874 76443-894 76443-914 76443-934 76443-954 76443-974 76443-994 76444-014 76444-034 76444-054 76444-074 76444-094 76444-524 76444-540 76444-556 76444-572 76444-592 76444-612 76444-632 76444-648 76444-664 76444-684 76444-704 76444-724 76444-744 76444-760 76444-776 76444-792 76444-812 76444-832 76444-852 76444-872 76444-888 76444-904 76444-920 76444-940 76444-960 76444-980 76445-000 76445-018 76445-038 76445-058 76445-422 76445-442 76445-462 76445-482 76445-502 76445-522 76445-542 76445-562 76445-582 76445-602 76445-622 76445-642 76445-662 76445-682 76445-702 76445-722 76445-742 76445-762 76445-782 76445-802 76445-822 76445-842 76445-862 76445-882 76445-902 76445-922 76445-942 76445-962 76445-982 76446-002 76446-022 76446-042 76446-152 76446-172 76446-192 76446-212 76446-232 76446-252 76446-272 76446-292 76446-312 76446-332 76446-352 76446-372 76446-392 76446-412 76446-432 76446-452 76446-472 76446-492 76446-512 76446-532 76446-552 76446-572 76446-592 76446-612 76446-632 76446-652 76446-672 76446-692 76446-712 76446-732
HALO® AQ-C18, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® AQ-C18 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å AQ-C18 columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for mixtures of polar and non-polar solutes
  • High retentivity of C18 with alternate selectivity
  • 100% aqueous compatible
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core',
More Product Information